Learn More
CELLTREAT Scientific Products 75cm2 Tissue Culture Flask - Vent Cap, Sterile
SDP
Catalog No. 50202080
Encompass_Preferred
Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Molded, interlocking T design for improved stacking on 75, 182, & 300cm2 and 250, 600 & 850mL size flasks. Vent caps feature 0.22¿m hydrophobic membrane.
Catalog No. 50-202-080 Supplier CELLTREAT Scientific Products Supplier No. 229341
Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Molded, interlocking T design for improved stacking on 75, 182, & 300cm2 and 250, 600 & 850mL size flasks. Vent caps feature 0.22¿m hydrophobic membrane.