Learn More
CELLTREAT Scientific Products 60mm x 15mm Tissue Culture Treated Dish w/Grip Ring, Sterile
SDP
Catalog No. 50202033
Encompass_Preferred
Cell Culture Dishes are available in Tissue Culture Treated and Non-treated Cell Culture versions. Non-treated Cell Culture Dishes have thicker and more durable plastic than standard Petri Dishes. 60, 70 and 100mm dishes available with a pronounced grip ring for improved handling. Vented lids contain molded spacers to allow gas exchange (except Part No.229655 which has a non-vented lid).
Catalog No. 50-202-033 Supplier CELLTREAT Scientific Products Supplier No. 229660
Cell Culture Dishes are available in Tissue Culture Treated and Non-treated Cell Culture versions. Non-treated Cell Culture Dishes have thicker and more durable plastic than standard Petri Dishes. 60, 70 and 100mm dishes available with a pronounced grip ring for improved handling. Vented lids contain molded spacers to allow gas exchange (except Part No.229655 which has a non-vented lid).