Learn More
Partec North America CELLTRIC 30MICRON NONSTRL250PK
Supplier:  Partec North America 0400422316
Encompass
CellTrics can be used to filter any kind of liquid suspension, e.g. for isolation of cells and nuclei from cell debris and aggregates and also for filtering either microorganisms or cell cultures.
Catalog No. NC9754599