Learn More
Partec North America CELLTRICS 150UM ST 50/PK
Catalog No. NC9021438
Encompass
CellTrics can be used to filter any kind of liquid suspension, e.g. for isolation of cells and nuclei from cell debris and aggregates and also for filtering either microorganisms or cell cultures.
Catalog No. NC9021438 Supplier Partec North America Supplier No. 040042329
CellTrics can be used to filter any kind of liquid suspension, e.g. for isolation of cells and nuclei from cell debris and aggregates and also for filtering either microorganisms or cell cultures.