Learn More
Bellco Glass, Inc. Celltrol Speed Control Box Tethered
SDP
Catalog No. 501946355
Encompass_Preferred
SST constructionDimensions 12 x 12 x 16"Digital tach input0-36 vdc; power @ 15a6' cable interface to CellTrol II. MADE IN THE USA
Catalog No. 50-194-6355 Supplier Bellco Glass, Inc. Supplier No. 780501917
SST constructionDimensions 12 x 12 x 16"Digital tach input0-36 vdc; power @ 15a6' cable interface to CellTrol II. MADE IN THE USA