Learn More
C LECTA GMBH CellTrypase,10x, 100mL, R&D
Catalog No. 5000600150
Encompass_Preferred
For research and process development using attachment-dependent cells. The enzyme enables gentle, reproducible dissociation while preserving cell viability and function. Ideal for adherent cell lines, stem and primary cells.- Purity: ≥ 95% (determined by HPLC)- Formulation: Liquid in PBS with EDTA, sterile filtered- Appearance: Clear to slightly colored solution- Storage: 2–8 °C- Delivery: 1–2 weeks (depending on region)- In-house ISO 9001-compliant manufacturing- Free from animal-derived raw materials and antibiotics- Produced with animal origin-free machinery- Net price excluding VAT and shipment costs
Catalog No. 50-006-00150 Supplier C LECTA GMBH Supplier No. 2210310X100
For research and process development using attachment-dependent cells. The enzyme enables gentle, reproducible dissociation while preserving cell viability and function. Ideal for adherent cell lines, stem and primary cells.- Purity: ≥ 95% (determined by HPLC)- Formulation: Liquid in PBS with EDTA, sterile filtered- Appearance: Clear to slightly colored solution- Storage: 2–8 °C- Delivery: 1–2 weeks (depending on region)- In-house ISO 9001-compliant manufacturing- Free from animal-derived raw materials and antibiotics- Produced with animal origin-free machinery- Net price excluding VAT and shipment costs