Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Spunbond Polypropylene Coveralls, Open Wrists and Ankles

Catalog No. 19133815 Shop All Cellucap Products
Change view
Click to view available options
Size:
Medium
Large
2X-Large
Size (English) Chest:
23.5 in.
25 in.
26.5 in.
Size (English) Inseam:
28 in.
29 in.
29.5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Chest Size (English) Inseam
19-133-815 Medium 23.5 in. 28 in.
19-133-817 2X-Large 26.5 in. 29.5 in.
19-133-816 Large 25 in. 29 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-133-815 Supplier Cellucap Supplier No. 55802M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls constructed of non-woven material offer lightweight splash protection from non-hazardous liquids and dry particulates.

  • Non-woven spunbond polypropylene
  • Lightweight splash protection from non-hazardous liquids and dry particulates
  • 1.25 oz. material
  • Open wrists and ankles
  • Collared

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Liquid, Particle
Model 55802
Size (English) Chest 23.5 in.
Size (English) Inseam 28 in.
Size (Metric) Chest 59.69 cm
Size (Metric) Inseam 71.12 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.