Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Lightweight Nylon Beard Net

Catalog No. 19046181 Shop All Cellucap Products
Change view
Click to view available options
Material:
Nylon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-046-181 Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-046-181 Supplier Cellucap Supplier No. BN5WT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight nylon beard net is made from a single strand, 15 denier nylon with 0.25 in. aperture.

  • Universal size
  • White
  • Nylon
  • 1,000/Cs.

Specifications

Material Nylon
Color White
Disposable Single-use
For Use With (Equipment) D903 Acrylic dispenser
Product Type General Purpose Beard Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.