Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Polypropylene Beard Restraint

Catalog No. 19046190 Shop All Cellucap Products
Change view
Click to view available options
Packaging Quantity:
1000/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-046-190 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-046-190 Supplier Cellucap Supplier No. BP4BL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Beard restraint helps to ensure a clean and hygienic work environment.

  • Polypropylene beard restraint
  • Serged seam style
  • Lightweight
  • Used in ISO 6/Class 1,000 environments

Specifications

Type Cleanroom Beard Cover
Size Regular
Material Polypropylene
Color Blue
Disposable Single-use
Latex Free Yes
Seam Type Serged
Certifications/Compliance ISO 6/Class 1,000
For Use With (Application) Cleanrooms, Pharmaceuticals
Packaging Quantity 1000/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.