Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Basic Spunbond Polypropylene Shoe Covers

Catalog No. 19314877 Shop All Cellucap Products
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-314-877 X-Large
19-262-125 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-314-877 Supplier Cellucap Supplier No. 28044
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Shoe covers provide lightweight protection from non-hazardous liquids and dry particulates. Ideal for visitors or light duty applications.

Specifications

Size X-Large
Material Spunbond Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Sole Type Plain
Model 28044
For Use With (Application) Ideal for visitors, light duty applications or general purpose coverage
Packaging Quantity 150 Pairs/Cs.
Packaging Type Dispenser Boxes
Resistance Fluid
Product Type Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.