Learn More
JNC AMERICA INC CELLUFINE MAX DEXS-VIRS,500ML
Supplier:  JNC AMERICA INC 21 802
Encompass
Cellufine MAX DexS is a new chromatography resin incorporating Dextran sulfate polymer surface modification. This non-animal derived pseudo affinity ligand can be used instead of immobilized Heparin for blood protein fractionation and viral capture.
Catalog No. NC2209501