Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology DRAQ7™ Cellular Dye, Far-Red Fluorescent, DNA Stain, 1 mL
SDP

Catalog No. 50194410
Encompass

DRAQ7™ is a far-red fluorescent DNA dye that specifically stains the nuclei of dead and permeabilized cells. This dye is highly versatile and compatible for use in combination with common labels such as GFP, FITC, and PE, making it an effective tool for multicolor flow cytometry applications.

  • Excitation range of 488-647 nm allows for flexible instrument settings and combination with standard fluorophores.
  • Specifically identifies dead or permeabilized cells by staining nuclear DNA.
  • Provided as a 0.3 mM stock solution for ease of use in experimental protocols.
  • Compatible with flow cytometry analysis workflows.
  • Store at 4°C, protected from light; do not freeze.

Catalog No. 50-194-410 Supplier Cell Signaling Technology Supplier No. 7406S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DRAQ7™ is a far-red fluorescent DNA dye that specifically stains the nuclei of dead and permeabilized cells. This dye is highly versatile and compatible for use in combination with common labels such as GFP, FITC, and PE, making it an effective tool for multicolor flow cytometry applications.

  • Excitation range of 488-647 nm allows for flexible instrument settings and combination with standard fluorophores.
  • Specifically identifies dead or permeabilized cells by staining nuclear DNA.
  • Provided as a 0.3 mM stock solution for ease of use in experimental protocols.
  • Compatible with flow cytometry analysis workflows.
  • Store at 4°C, protected from light; do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.