Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cellular retinoic acid-binding protein 1 (human) | >95.0% | 10 UG
SDP

Catalog No. 5000290538
Encompass_Preferred

Recombinant human cellular retinoic acid-binding protein 1 (CRABP1), expressed in E. coli and provided tag-free, is supplied for research applications involving retinoic acid signaling, protein interaction studies, and biochemical assays. The purified protein (~14 kDa) is offered in small quantities suitable for binding studies and method development.

  • Recombinant human sequence for relevant biological activity.
  • Tag free to minimize interference in functional assays.
  • Expressed in E. coli for efficient production and consistency.
  • Approximate molecular weight 14 kDa for identification and QC.
  • Available in small sizes such as 10 μg for low-consumption experiments.
  • Purity greater than 95% to support biochemical and binding studies.

Catalog No. 50-002-90538 Supplier Medchemexpress LLC Supplier No. HYP7568710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human cellular retinoic acid-binding protein 1 (CRABP1), expressed in E. coli and provided tag-free, is supplied for research applications involving retinoic acid signaling, protein interaction studies, and biochemical assays. The purified protein (~14 kDa) is offered in small quantities suitable for binding studies and method development.

  • Recombinant human sequence for relevant biological activity.
  • Tag free to minimize interference in functional assays.
  • Expressed in E. coli for efficient production and consistency.
  • Approximate molecular weight 14 kDa for identification and QC.
  • Available in small sizes such as 10 μg for low-consumption experiments.
  • Purity greater than 95% to support biochemical and binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.