Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cellular retinoic acid-binding protein 1, human recombinant | >95.0% | 50 UG
SDP

Catalog No. 5000290135
Encompass_Preferred

Recombinant human cellular retinoic acid-binding protein 1 (CRABP1), expressed in E. coli and supplied tag-free as a lyophilized powder. Supplied as a 50 μg unit, the protein is >95% pure, ~14 kDa, full length (M1-E137), formulated in 50 mM Tris-HCl, 300 mM NaCl, pH 8.0, 10% glycerol, with endotoxin <1 EU/μg. Store at -20°C; reconstitute to at least 100 μg/mL in ddH2O and follow laboratory handling procedures.

  • Recombinant human protein expressed in E. coli.
  • Tag free preparation suitable for functional assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight of 14 kDa.
  • Lyophilized powder formulation for long-term storage.
  • Low endotoxin level (<1 EU/μg) for sensitive applications.
  • Reconstitution recommended to at least 100 μg/mL in ddH2O.
  • Stable at 4°C for 1 week after reconstitution or at -20°C for long-term storage with carrier protein.

Catalog No. 50-002-90135 Supplier Medchemexpress LLC Supplier No. HYP7568750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human cellular retinoic acid-binding protein 1 (CRABP1), expressed in E. coli and supplied tag-free as a lyophilized powder. Supplied as a 50 μg unit, the protein is >95% pure, ~14 kDa, full length (M1-E137), formulated in 50 mM Tris-HCl, 300 mM NaCl, pH 8.0, 10% glycerol, with endotoxin <1 EU/μg. Store at -20°C; reconstitute to at least 100 μg/mL in ddH2O and follow laboratory handling procedures.

  • Recombinant human protein expressed in E. coli.
  • Tag free preparation suitable for functional assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight of 14 kDa.
  • Lyophilized powder formulation for long-term storage.
  • Low endotoxin level (<1 EU/μg) for sensitive applications.
  • Reconstitution recommended to at least 100 μg/mL in ddH2O.
  • Stable at 4°C for 1 week after reconstitution or at -20°C for long-term storage with carrier protein.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.