Learn More
Medchemexpress LLC Cellular retinoic acid-binding protein 1, recombinant (human) | >95.0% | 100 UG
SDP
Catalog No. 5000290537
Encompass_Preferred
Recombinant human cellular retinoic acid-binding protein 1 supplied as a lyophilized, tag-free protein for biochemical and cell biology applications. Expressed in E. coli and purified to >95% purity, the protein is provided as 100 μg and is suitable for assays requiring native CRABP1 activity or antibody validation. Follow supplier reconstitution and storage guidance for optimal stability.
Catalog No. 50-002-90537 Supplier Medchemexpress LLC Supplier No. HYP75687100UG
Recombinant human cellular retinoic acid-binding protein 1 supplied as a lyophilized, tag-free protein for biochemical and cell biology applications. Expressed in E. coli and purified to >95% purity, the protein is provided as 100 μg and is suitable for assays requiring native CRABP1 activity or antibody validation. Follow supplier reconstitution and storage guidance for optimal stability.