Learn More
Medchemexpress LLC Cellular retinoic acid-binding protein 2 (CRABP2) | 100 UG
SDP
Catalog No. 5000290539
Encompass_Preferred
Recombinant human cellular retinoic acid-binding protein 2 (CRABP2), expressed in E. coli and supplied as a His-tagged, lyophilized protein. Supplied as a 100 μg vial in lyophilized powder form, formulated from Tris-HCl and NaCl. Suitable for in vitro biochemical and cell biology studies of retinoic acid binding and signaling.
Catalog No. 50-002-90539 Supplier Medchemexpress LLC Supplier No. HYP75688100UG
Recombinant human cellular retinoic acid-binding protein 2 (CRABP2), expressed in E. coli and supplied as a His-tagged, lyophilized protein. Supplied as a 100 μg vial in lyophilized powder form, formulated from Tris-HCl and NaCl. Suitable for in vitro biochemical and cell biology studies of retinoic acid binding and signaling.