Learn More
Dojindo Molecular Technologies Inc Cellular Senescence Plate Assay Kit (100TESTS), easily quantify SA-β-gal activity via plate reader, suitable for multiple sample assays
SDP
Catalog No. NC1608115
Encompass
This product is a simple detection kit by plate assay for senescence-associated beta-galactosidase (SA-β-gal) activity which is used as a marker for senescent cells. By simply adding SPiDER-Gal, a reagent for detection of β-galactosidase, to 96 well plates, this kit allows you to quantify SA-β-gal activity and makes it possible to evaluate multiple specimens. When normalization is done by the results obtained by counting cells, quantifying nucleic acids, or quantifying proteins, the measured values obtained using this kit become available for evaluating SA-β-gal activity according to cell number. The features of cellular senescence were summarized based on modern publications that have a high number of citations on this page (https://www.dojindo.com/senescent-cell-detection-selection-guide).
Catalog No. NC1608115 Supplier Dojindo Molecular Technologies Inc Supplier No. SG0505
This product is a simple detection kit by plate assay for senescence-associated beta-galactosidase (SA-β-gal) activity which is used as a marker for senescent cells. By simply adding SPiDER-Gal, a reagent for detection of β-galactosidase, to 96 well plates, this kit allows you to quantify SA-β-gal activity and makes it possible to evaluate multiple specimens. When normalization is done by the results obtained by counting cells, quantifying nucleic acids, or quantifying proteins, the measured values obtained using this kit become available for evaluating SA-β-gal activity according to cell number. The features of cellular senescence were summarized based on modern publications that have a high number of citations on this page (https://www.dojindo.com/senescent-cell-detection-selection-guide).