Learn More
Cell Biolabs Inc Cellular Senescence Detection Kit (SA-Beta-Gal Staining), 50 assays
Catalog No. NC9849906
Encompass_Preferred
Senescent cells show common biochemical markers such as expression of an acidic senescence-associated ß-galactosidase (SA-ß-Gal) activity. While senescence has been characterized primarily in cultured cells, there is also evidence that it occurs in vivo. Cells expressing markers of senescence such as SA-ß-Gal have been identified in normal tissues. Cell Biolabs' Cellular Senescence Assay Kit provides an easy-to-use and efficient method to determine cellular senescence. SA-β-galactosidase catalyzes the hydrolysis of X-gal, which produces a blue color. Each kit provides sufficient quantities to perform up to 50 assays in 35 mm wells.
Catalog No. NC9849906 Supplier Cell Biolabs Inc Supplier No. CBA230
Senescent cells show common biochemical markers such as expression of an acidic senescence-associated ß-galactosidase (SA-ß-Gal) activity. While senescence has been characterized primarily in cultured cells, there is also evidence that it occurs in vivo. Cells expressing markers of senescence such as SA-ß-Gal have been identified in normal tissues. Cell Biolabs' Cellular Senescence Assay Kit provides an easy-to-use and efficient method to determine cellular senescence. SA-β-galactosidase catalyzes the hydrolysis of X-gal, which produces a blue color. Each kit provides sufficient quantities to perform up to 50 assays in 35 mm wells.