Learn More
Cell Biolabs Inc 96-Well Cellular Senescence Assay Kit (SA-Beta-Gal Activity, Fluorometric), 120 assays
Catalog No. NC9780342
Encompass_Preferred
Senescent cells show common biochemical markers such as expression of an acidic senescence-associated ß-galactosidase (SA-ß-Gal) activity. While senescence has been characterized primarily in cultured cells, there is also evidence that it occurs in vivo. Cells expressing markers of senescence such as SA-ß-Gal have been identified in normal tissues. Cell Biolabs' 96-well Cellular Senescence Assay Kit provides an easy-to-use and efficient method to determine the cellular senescence by measuring SA-ß-Gal activity using a fluorometric substrate. This quantitative assay uses cell lysate for both SA-β-galactosidase activity determination and normalization of samples containing different cell numbers. Each kit provides sufficient quantities to perform up to 120 assays in a 96-well plate.
Catalog No. NC9780342 Supplier Cell Biolabs Inc Supplier No. CBA231
Senescent cells show common biochemical markers such as expression of an acidic senescence-associated ß-galactosidase (SA-ß-Gal) activity. While senescence has been characterized primarily in cultured cells, there is also evidence that it occurs in vivo. Cells expressing markers of senescence such as SA-ß-Gal have been identified in normal tissues. Cell Biolabs' 96-well Cellular Senescence Assay Kit provides an easy-to-use and efficient method to determine the cellular senescence by measuring SA-ß-Gal activity using a fluorometric substrate. This quantitative assay uses cell lysate for both SA-β-galactosidase activity determination and normalization of samples containing different cell numbers. Each kit provides sufficient quantities to perform up to 120 assays in a 96-well plate.