Learn More
Enzo Life Sciences Cellular senescence live cell analysis assay (1x10assays)
SDP
Catalog No. NC0988954
Encompass_Preferred
Simple assay to quantify senescence-associated β-galactosidase activity in live cells using flow cytometry. Assay kit that uses a fluorogenic substrate to measure SA-β-gal activity in live cells. Applications: Flow Cytometry. Contents: Cell pretreatment solution (1000x). SA-β-gal Substrate (200x)
Catalog No. NC0988954 Supplier Enzo Life Sciences Supplier No. ENZKIT1300010
Simple assay to quantify senescence-associated β-galactosidase activity in live cells using flow cytometry. Assay kit that uses a fluorogenic substrate to measure SA-β-gal activity in live cells. Applications: Flow Cytometry. Contents: Cell pretreatment solution (1000x). SA-β-gal Substrate (200x)