Learn More
Medchemexpress LLC HY-B2220 1g Medchemexpress, Cellulase CAS:9012-54-8 Purity:>98%
SDP
Catalog No. 501933113
Encompass_Preferred
Medchemexpress, HY-B2220 1g Cellulase CAS:9012-54-8 Cellulase is an enzyme catalyzing the hydrolysis of certain linkages in cellulose and other carbohydrates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3113 Supplier Medchemexpress LLC Supplier No. HYB22201G
Medchemexpress, HY-B2220 1g Cellulase CAS:9012-54-8 Cellulase is an enzyme catalyzing the hydrolysis of certain linkages in cellulose and other carbohydrates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.