Learn More
Medchemexpress LLC Cellulase | 9012-54-8 | 500 MG
SDP
Catalog No. 5000409715
Encompass_Preferred
Cellulase is an enzyme that catalyzes the hydrolysis of specific linkages in cellulose and other carbohydrates. It is intended for research use only.
  • Exhibits significant antibiofilm activity
  • Can increase Capillary Suction Time (CST) in activated sludge when combined with protease
  • Decreases the particle size of activated sludge
Catalog No. 50-004-09715 Supplier Medchemexpress LLC Supplier No. HYB2220500MG
Cellulase is an enzyme that catalyzes the hydrolysis of specific linkages in cellulose and other carbohydrates. It is intended for research use only.
  • Exhibits significant antibiofilm activity
  • Can increase Capillary Suction Time (CST) in activated sludge when combined with protease
  • Decreases the particle size of activated sludge