Learn More
Bioworld Cellulase enzyme (From Trichoderma), 5 g
Catalog No. NC0999878
Encompass_Preferred
Cellulase (EC 3.2.1.4) is an enzyme used to isolate intact cells and protoplasts from plant tissue that is derived from Trichoderma viride. Reaction: Hydrolyzes 1,4-glucosidic linkages in cellulose, lichenin and cereal beta-D-glucans. Activity : 150,000 CU/g Synonyms: endo-1,4-beta-glucanase, carboxymethyl cellulase (CMCase), endo-1,4-beta-D-glucanase, beta-1,4-glucanase, beta-1,4-endoglucan hydrolase, and celludextrinase
Catalog No. NC0999878 Supplier Bioworld Supplier No. 215000031
Cellulase (EC 3.2.1.4) is an enzyme used to isolate intact cells and protoplasts from plant tissue that is derived from Trichoderma viride. Reaction: Hydrolyzes 1,4-glucosidic linkages in cellulose, lichenin and cereal beta-D-glucans. Activity : 150,000 CU/g Synonyms: endo-1,4-beta-glucanase, carboxymethyl cellulase (CMCase), endo-1,4-beta-D-glucanase, beta-1,4-glucanase, beta-1,4-endoglucan hydrolase, and celludextrinase