Learn More
Sigma Aldrich Fine Chemicals Biosciences Cellulase enzyme blend
Catalog No. 502898729
Encompass
This product contains cellulases AY-glucosidases and hemicellulase for the application of degrading cellulose to fermentable sugars. This product is effective on a wide variety of pre-treated lignocellulosic bimass materials for converting the carbohydrates in these materials into simple sugars prior to fermentation for application in biofuels research.
Catalog No. 50-289-8729 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE002050ML
This product contains cellulases AY-glucosidases and hemicellulase for the application of degrading cellulose to fermentable sugars. This product is effective on a wide variety of pre-treated lignocellulosic bimass materials for converting the carbohydrates in these materials into simple sugars prior to fermentation for application in biofuels research.