Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Cellulase [Onozuka R-10], 5 Grams
SDP

Catalog No. 50213230
Encompass_Preferred

From Trichadema Viride. Enzyme mixture often used in combination with Macerozyme R-10 for isolation of protoplasts. One unit of Cellulase will liberate 1.0 µmole of glucose from carboxymethyl cellulose. Optimum pH range: 4-5°

Ingredients:

Cellulase: 1.0 U/mg
Pectinase: 0.4 U/mg
Hemicellulase: 1.0 U/mg
alpha-Amylase: 0.6 U/mg
Protease: 0.01 U/mg

Catalog No. 50-213-230 Supplier Research Products International Corp Supplier No. C322005.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

From Trichadema Viride. Enzyme mixture often used in combination with Macerozyme R-10 for isolation of protoplasts. One unit of Cellulase will liberate 1.0 µmole of glucose from carboxymethyl cellulose. Optimum pH range: 4-5°

Ingredients:

Cellulase: 1.0 U/mg
Pectinase: 0.4 U/mg
Hemicellulase: 1.0 U/mg
alpha-Amylase: 0.6 U/mg
Protease: 0.01 U/mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.