Learn More
Research Products International Corp Cellulase RS [Onozuka RS], 10 Grams
SDP
Catalog No. 50213232
Encompass_Preferred
From mutant Trichoderma viride derived from the parent strain for Cellulase Onozuka R-10. Cellulase RS contains a high activity of decomposing natural celluloses. Can be used to obtain protoplasts and dissolve cell walls in a wide range of plants.
Catalog No. 50-213-232 Supplier Research Products International Corp Supplier No. C3240010.0
From mutant Trichoderma viride derived from the parent strain for Cellulase Onozuka R-10. Cellulase RS contains a high activity of decomposing natural celluloses. Can be used to obtain protoplasts and dissolve cell walls in a wide range of plants.