Learn More
Research Products International Corp Cellulase RS [Onozuka RS], 5 Grams
SDP
Catalog No. 50213233
Encompass_Preferred
From mutant Trichoderma viride derived from the parent strain for Cellulase Onozuka R-10. Cellulase RS contains a high activity of decomposing natural celluloses. Can be used to obtain protoplasts and dissolve cell walls in a wide range of plants.
Catalog No. 50-213-233 Supplier Research Products International Corp Supplier No. C324005.0
From mutant Trichoderma viride derived from the parent strain for Cellulase Onozuka R-10. Cellulase RS contains a high activity of decomposing natural celluloses. Can be used to obtain protoplasts and dissolve cell walls in a wide range of plants.