Learn More
Bioworld Cellulase RS [Onozuka RS], 5 g
Catalog No. 50198965
Encompass_Preferred
From mutant Trichoderma viride derived from the parent strain for Cellulase Onozuka R-10. Cellulase RS contains a high activity of decomposing natural celluloses. Can be used to obtain protoplasts and dissolve cell walls in a wide range of plants.* Soluble in water* Optimum pH range 4.0-5.0* Optimum temperature range 50° to 60°CUsually ships within 24 hours.This product is sold for laboratory research use only.
Catalog No. 50-198-965 Supplier Bioworld Supplier No. 215600052
From mutant Trichoderma viride derived from the parent strain for Cellulase Onozuka R-10. Cellulase RS contains a high activity of decomposing natural celluloses. Can be used to obtain protoplasts and dissolve cell walls in a wide range of plants.* Soluble in water* Optimum pH range 4.0-5.0* Optimum temperature range 50° to 60°CUsually ships within 24 hours.This product is sold for laboratory research use only.