Learn More
Bioworld Cellulase (Onozuka R-10), 5 g
Catalog No. 50198963
Encompass_Preferred
Cellulase (Onozuka R-10) From Trichadema Viride. Enzyme mixture often used in combination with Macerozyme R-10 (M22010) for isolation of plant protoplasts. One unit of Cellulase will liberate 1.0 µmole of glucose from carboxymethyl cellulose.Optimum pH range: 4-5.Soluble in water (10mg/ml)Contents:Cellulase: 1.0 U/mg Pectinase: 0.4 U/mg Hemicellulase: 1.0 U/mg alpha-Amylase: 0.6 U/mg Protease: 0.01 U/mg
Catalog No. 50-198-963 Supplier Bioworld Supplier No. 215600032
Cellulase (Onozuka R-10) From Trichadema Viride. Enzyme mixture often used in combination with Macerozyme R-10 (M22010) for isolation of plant protoplasts. One unit of Cellulase will liberate 1.0 µmole of glucose from carboxymethyl cellulose.Optimum pH range: 4-5.Soluble in water (10mg/ml)Contents:Cellulase: 1.0 U/mg Pectinase: 0.4 U/mg Hemicellulase: 1.0 U/mg alpha-Amylase: 0.6 U/mg Protease: 0.01 U/mg