Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cellulose acetate

Catalog No. 18095525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-095-525G 25 g
18-095-5500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-095-525G Supplier Sigma Aldrich Supplier No. 18095525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cellulose is a naturally occurring biodegradable and biocompatible polymer. Cellulose based polymers find wide variety of applications as base matrix in biopharmaceutical industry. Cellulose acetate is soluble in acetone. Cellulose acetate can be easily electrospun into its nanofibers.Cellulose acetate was used in the fabrication of gold microelectrode for electrochemical monochloramine measurement. Cellulose acetate based membranes are used in the separations in aqueous systems and in reverse osmosis process. It has been reported to be used to desalinate seawater.Cellulose acetate was used for biosensor encapsulation. It was used in the preparation of cellulose acetate nanofibre felt structure, cellulose acetate fibers.

Specifications

CAS 9004-35-7
Density 1.3 g/mL (at 25°C)
Refractive Index n20/D 1.475 (literature)
MDL Number MFCD00081496
Quantity 25 g
Synonym Acetylcellulose
Molecular Weight (g/mol) 30000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.