Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cellulose acetate

Catalog No. 41902825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-902-825G 25 g
41-902-8500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-902-825G Supplier Sigma Aldrich Supplier No. 41902825G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cellulose acetate (CA) is a class of cellulose based esters which can be produced by a reaction of cellulose with acetic acid and acetic anhydride. It can be used in a variety of forms such as films, membranes, and fibers. It can be used as a biodegradable polymer in medical and automotive applications.CA can be used as a material in packaging, clothing and pharmaceuticals industries. It can also be used in the formation of polymeric electrolyte for the fabrication of electrochemical devices like fuel cells, solar cells and supercapacitors.

Specifications

CAS 9004-35-7
Density 1.3 g/mL (at 25°C)
Refractive Index n20/D 1.475 (literature)
Quantity 25 g
MDL Number MFCD00081496
Synonym Acetylcellulose
Molecular Weight (g/mol) 50000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.