Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellulose acetate phthalate , MilliporeSigma™ Supelco™

Catalog No. 111015801 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5801 100 g
11-101-5800 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5801 Supplier MilliporeSigma Supelco Supplier No. 22192100GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cellulose acetate phthalate (CAP) is a microbicide which inactivates sexually transmitted disease (STD) pathogens like HIV-1. CAP has been used for enteric film coating of tablets and capsules.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00072769

Specifications

CAS 9004-38-0
Packaging Poly Bottle
Linear Formula C116H116O64
Quantity 100 g
Formula Weight 2534.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.