Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellulose Acetate Trimellitate, Thermo Scientific™

Catalog No. AC404305000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404305000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404305000 Supplier Thermo Scientific Chemicals Supplier No. 404305000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52907-01-4
Molecular Formula C164H174O111
Molecular Weight (g/mol) 3921.085
InChI Key WZNRVWBKYDHTKI-UHFFFAOYSA-N
Synonym cellulose acetate trimellitate, cellulose, acetate 1,2,4-benzenetricarboxylate
PubChem CID 16132304
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C.CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C.C1=CC(=C(C=C1C(=O)O)C(=O)O)C(=O)OCC2C(C(C(C(O2)OC(=O)C3=C(C=C(C=C3)C(=O

Specifications

Color White
Physical Form Fine Crystalline Powder
Merck Index 10, 1926
Quantity 500 g
Infrared Spectrum Authentic
Chemical Name or Material Cellulose acetate trimellitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.