Learn More
Medchemexpress LLC Cellulose, carboxymethyl ether | 9000-11-7 | <5% | 10 G
SDP
Catalog No. 5000409686
Encompass_Preferred
CM Cellulose CM-32, a cellulose derivative, finds extensive application in biomedical fields such as tissue engineering, wound dressing, and the creation of 3D-scaffolds for biocompatible implants and artificial organs. It can also be utilized in the diagnosis of various diseases and has been shown to decrease osteoclast formation. The compound is suitable for life science-related research as a biological material or organic compound.
  • Supports adhesion and proliferation of mouse fibroblasts
  • Significantly reduces osteoclast formation in a murine bone marrow model
  • Lowers bone resorption in bone marrow cells
  • Increases formazan formation by osteoclast progenitors
  • Enables synthesis of separate bundles of cellulose microfibrils
Catalog No. 50-004-09686 Supplier Medchemexpress LLC Supplier No. HYB2221C10G
CM Cellulose CM-32, a cellulose derivative, finds extensive application in biomedical fields such as tissue engineering, wound dressing, and the creation of 3D-scaffolds for biocompatible implants and artificial organs. It can also be utilized in the diagnosis of various diseases and has been shown to decrease osteoclast formation. The compound is suitable for life science-related research as a biological material or organic compound.
  • Supports adhesion and proliferation of mouse fibroblasts
  • Significantly reduces osteoclast formation in a murine bone marrow model
  • Lowers bone resorption in bone marrow cells
  • Increases formazan formation by osteoclast progenitors
  • Enables synthesis of separate bundles of cellulose microfibrils