Learn More
Medchemexpress LLC Cellulose, carboxymethyl ether, sodium salt | 9004-32-4 | 81472 | 100mg
SDP
Catalog No. 5000266136
Encompass_Preferred
Sodium carboxymethyl cellulose (CMC-Na) (MW 250000) is a sodium salt of carboxymethyl cellulose Sodium carboxymethyl cellulose has adsorption and corrosion inhibition on low-carbon steel in an acidic medium Sodium carboxymethyl cellulose can be used as a thickener paste and barrier agent[1][2][3]
Catalog No. 50-002-66136 Supplier Medchemexpress LLC Supplier No. HYY1889A100MG
Add to Cart
Add to Cart
Sodium carboxymethyl cellulose (CMC-Na) (MW 250000) is a sodium salt of carboxymethyl cellulose Sodium carboxymethyl cellulose has adsorption and corrosion inhibition on low-carbon steel in an acidic medium Sodium carboxymethyl cellulose can be used as a thickener paste and barrier agent[1][2][3]