Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellulose DFS-0, Microcrystalline, For thin layer chromatography, MilliporeSigma™ Supelco™

Catalog No. 111015795 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5795 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5795 Supplier MilliporeSigma Supelco Supplier No. 099061KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00081512
Synonym Cellulose powder; Cotton linters

Specifications

Physical Form MicroCrystalline
CAS 9004-34-6
Quantity 1 kg
Grade TLC
RTECSNumber FJ5691460
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.