Learn More
Medchemexpress LLC Cellulose, methyl ether | 9004-67-5 | MFCD00081763 | [C6H7O2(OH)x(OCH3)y]n | 25G
SDP
Catalog No. 5000235595
Encompass_Preferred
Methyl cellulose (viscosity 4000 mPa·s) is a non-ionic cellulose ether supplied as a white to off-white solid. It acts as a thickener, stabilizer, film former, and thermogelation agent for aqueous systems used in laboratory, pharmaceutical, cosmetic, food, and industrial formulations. It modifies rheology and provides reversible gelling on heating, making it useful for formulation and processing control.
  • Used as a thickener and stabilizer in aqueous formulations.
  • Provides thermogelation and reversible gelling on heating.
  • White to off-white powder physical form.
  • Available in laboratory pack sizes for small-scale use.
  • Suitable as a pharmaceutical excipient and cosmetic additive.
Catalog No. 50-002-35595 Supplier Medchemexpress LLC Supplier No. HY125861E25G
Methyl cellulose (viscosity 4000 mPa·s) is a non-ionic cellulose ether supplied as a white to off-white solid. It acts as a thickener, stabilizer, film former, and thermogelation agent for aqueous systems used in laboratory, pharmaceutical, cosmetic, food, and industrial formulations. It modifies rheology and provides reversible gelling on heating, making it useful for formulation and processing control.
  • Used as a thickener and stabilizer in aqueous formulations.
  • Provides thermogelation and reversible gelling on heating.
  • White to off-white powder physical form.
  • Available in laboratory pack sizes for small-scale use.
  • Suitable as a pharmaceutical excipient and cosmetic additive.