Learn More
Medchemexpress LLC Cellulose, methyl ether | 9004-67-5 | MFCD00081763 | C20H38O11 | 500G
SDP
Catalog No. 5000234952
Encompass_Preferred
Methyl cellulose (viscosity: 4000 mPa·s) is a non-ionic cellulose ether supplied as a white powder. It acts as a thickener, stabilizer, emulsifier, and thermogelation agent for formulation and research applications. The product is offered in multiple pack sizes (including 500 g) and is provided with a certificate of analysis, safety data sheet, and handling documentation. Intended for research use only.
  • Non-ionic cellulose ether with thermogelation properties.
  • Viscosity 4000 mPa·s (typical), suitable for thickening and stabilizing.
  • Functions as an emulsifier and film former in formulations.
  • Supplied as a powder with certificate of analysis and safety data sheet available.
  • Available in multiple pack sizes including 500 g.
  • For research use only; follow handling instructions in SDS.
Catalog No. 50-002-34952 Supplier Medchemexpress LLC Supplier No. HY125861E500G
Methyl cellulose (viscosity: 4000 mPa·s) is a non-ionic cellulose ether supplied as a white powder. It acts as a thickener, stabilizer, emulsifier, and thermogelation agent for formulation and research applications. The product is offered in multiple pack sizes (including 500 g) and is provided with a certificate of analysis, safety data sheet, and handling documentation. Intended for research use only.
  • Non-ionic cellulose ether with thermogelation properties.
  • Viscosity 4000 mPa·s (typical), suitable for thickening and stabilizing.
  • Functions as an emulsifier and film former in formulations.
  • Supplied as a powder with certificate of analysis and safety data sheet available.
  • Available in multiple pack sizes including 500 g.
  • For research use only; follow handling instructions in SDS.