Learn More
Medchemexpress LLC Cellulose, methyl ether | 9004-67-5 | MFCD00081763 | Not specified | Not defined (polymeric material; molecular weight varies by viscosity grade) | No fixed formula (polymeric cellulose methyl ether) | 50G
SDP
Catalog No. 5000235596
Encompass_Preferred
Methyl cellulose (viscosity 4000 mPa·s) is a non-ionic cellulose ether supplied for research use as a white to off-white powder. It functions as a water-soluble polymeric thickener and rheology modifier that gels on heating and is commonly used in laboratory formulations for stabilization, emulsification, binding, and viscosity control.
  • Provides viscosity control and thermogelation properties.
  • Water soluble and non-ionic polymer.
  • Used as a thickener, stabilizer, emulsifier, and binder.
  • Available in laboratory pack sizes, including 50 g.
  • For research use only; no analytical purity percentage specified.
Catalog No. 50-002-35596 Supplier Medchemexpress LLC Supplier No. HY125861E50G
Methyl cellulose (viscosity 4000 mPa·s) is a non-ionic cellulose ether supplied for research use as a white to off-white powder. It functions as a water-soluble polymeric thickener and rheology modifier that gels on heating and is commonly used in laboratory formulations for stabilization, emulsification, binding, and viscosity control.
  • Provides viscosity control and thermogelation properties.
  • Water soluble and non-ionic polymer.
  • Used as a thickener, stabilizer, emulsifier, and binder.
  • Available in laboratory pack sizes, including 50 g.
  • For research use only; no analytical purity percentage specified.