Learn More
Medchemexpress LLC Cellulose, methyl ether | 9004-67-5 | MFCD00081763 | 98.0% | Not specified (polymeric) | [C6H7O2(OH)x(OCH3)y]n | 100G
SDP
Catalog No. 5000235963
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether powder specified at a viscosity of 40,000 mPa·s and a purity of 98.0%. It exhibits surface-active and thermogelation properties and is used as a thickener, stabilizer, emulsifier, and formulation excipient in pharmaceutical, food, cosmetic, and construction applications.
  • High viscosity grade 40,000 mPa·s for strong thickening.
  • Thermogelation behavior enables temperature-responsive gel formation.
  • Non-ionic, water-soluble polymer compatible with many formulation types.
  • Useful as a thickener, stabilizer, emulsifier, and drug-delivery excipient.
  • Available in laboratory pack sizes including 100 g.
  • Typical supplied purity 98.0% ensures consistent performance.
Catalog No. 50-002-35963 Supplier Medchemexpress LLC Supplier No. HY125861D100G
Methyl cellulose is a non-ionic cellulose ether powder specified at a viscosity of 40,000 mPa·s and a purity of 98.0%. It exhibits surface-active and thermogelation properties and is used as a thickener, stabilizer, emulsifier, and formulation excipient in pharmaceutical, food, cosmetic, and construction applications.
  • High viscosity grade 40,000 mPa·s for strong thickening.
  • Thermogelation behavior enables temperature-responsive gel formation.
  • Non-ionic, water-soluble polymer compatible with many formulation types.
  • Useful as a thickener, stabilizer, emulsifier, and drug-delivery excipient.
  • Available in laboratory pack sizes including 100 g.
  • Typical supplied purity 98.0% ensures consistent performance.