Learn More
Crescent Chemical Co Inc CELLULOSE MICROCRYSTALLINE
SDP
Catalog No. 50247328
Encompass_Preferred
Cellulose microcrystalline ca. 0.020 mm research grade Suitable for TLC, no binder required. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 14205.02 CAS 9004-34-6
Catalog No. 50-247-328 Supplier Crescent Chemical Co Inc Supplier No. SE14205.02
Cellulose microcrystalline ca. 0.020 mm research grade Suitable for TLC, no binder required. Size - 1KG Storage Conditions - +15 °C TO +30 °C Catalog Number - 14205.02 CAS 9004-34-6