Learn More
Crescent Chemical Co Inc CELLULOSE ;ONOZUKA R-10+
SDP
Supplier:  Crescent Chemical Co Inc SE16419.02
Encompass_Preferred
Cellulase »Onozuka R-10« from Trichoderma viride ca. 1 U/mg A multi-component enzyme system (1). Although the preparation has high cellulase activity, it still contains hemicellulases, and it degrades mannans, xylans, galactomannans, pectins and other polysaccharides. Widely used for the isolation of prot
Catalog No. 50-247-412