Learn More
Crescent Chemical Co Inc CELLULOSE ;ONOZUKA RS+
SDP
Catalog No. 50247416
Encompass_Preferred
Cellulase »Onozuka RS« from Trichoderma viride ca. 2 U/mg A multi-component enzyme system (1). Although the preparation has high cellulase activity (EC 3.2.1.4), it still contains hemicellulases, and it degrades mannans, xylans, galactomannans, pectins and other polysaccharides. Contains about three tim
Catalog No. 50-247-416 Supplier Crescent Chemical Co Inc Supplier No. SE16420.02
Cellulase »Onozuka RS« from Trichoderma viride ca. 2 U/mg A multi-component enzyme system (1). Although the preparation has high cellulase activity (EC 3.2.1.4), it still contains hemicellulases, and it degrades mannans, xylans, galactomannans, pectins and other polysaccharides. Contains about three tim