Learn More
Crescent Chemical Co Inc CELLULOSE ;ONOZUKA RS+
SDP
Supplier:  Crescent Chemical Co Inc SE16420.01
Encompass_Preferred
Cellulase »Onozuka RS« from Trichoderma viride ca. 2 U/mg A multi-component enzyme system (1). Although the preparation has high cellulase activity (EC 3.2.1.4), it still contains hemicellulases, and it degrades mannans, xylans, galactomannans, pectins and other polysaccharides. Contains about three tim
Catalog No. 50-247-415