Learn More
Analytical Sales & Services Cellvis 96-well Black Polystyrene/Glass Bottom Plate with high performance #1.5 cover glass
Catalog No. 5000807010
Encompass
96-well Cell Culture/Imaging Microplate\n\n• Black polystyrene frame with high performance #1.5 cover glass (0.170±0.005mm)\n• Flat glass bottom\n• Suitable for long term tissue culture\n• Sterilized by Gamma Irradiation and individually packed\n\nQty/Pkg: 20
Catalog No. 50-008-07010 Supplier Analytical Sales & Services Supplier No. 961515
96-well Cell Culture/Imaging Microplate\n\n• Black polystyrene frame with high performance #1.5 cover glass (0.170±0.005mm)\n• Flat glass bottom\n• Suitable for long term tissue culture\n• Sterilized by Gamma Irradiation and individually packed\n\nQty/Pkg: 20