Learn More
STEMBIOSYS INC CELLvo™ Matrix, T-150 Flask, cell growth area 150 cm2, working volume 30-45 mL, canted and vented, polystyrene material, 5 flasks per sleeve
SDP
Catalog No. NC1677201
Encompass_Preferred
CELLvo™ Matrix T-150 Flasks are a ready-to-use cell culture substrate which provides a native microenvironment that aims to replicate the biochemical, structural and mechanical cues of the native niche where cells naturally reside and proliferate. The matrix is composed of over 150 proteins that were secreted and assembled by human stromal cells during production. The final product is cell free with only the ECM attached to the surface of the culture vessel. CELLvo™ Matrix can be used for rapid expansion of high quality MSCs and other adherent cells.
Catalog No. NC1677201 Supplier STEMBIOSYS INC Supplier No. BMHPMET150
CELLvo™ Matrix T-150 Flasks are a ready-to-use cell culture substrate which provides a native microenvironment that aims to replicate the biochemical, structural and mechanical cues of the native niche where cells naturally reside and proliferate. The matrix is composed of over 150 proteins that were secreted and assembled by human stromal cells during production. The final product is cell free with only the ECM attached to the surface of the culture vessel. CELLvo™ Matrix can be used for rapid expansion of high quality MSCs and other adherent cells.