Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLytcTM M Cell Lyss Re250ML
Catalog No. 502905579
Encompass
CelLytic M is a proprietary detergent SOLUTIONon designed for efficient whole-cell protein extraction from cultured mammalian cells. It enables efficient and rapid cell lysis and solubilization of proteins for both suspension and adherent cells. Lysates can be used in many downstream applications without removing the CelLytic M such as reporter gene assays Western blots/immunoprecipitation electrophoretic mobility shift assays phosphatase assays and kinase asssays.
Catalog No. 50-290-5579 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C2978250ML
CelLytic M is a proprietary detergent SOLUTIONon designed for efficient whole-cell protein extraction from cultured mammalian cells. It enables efficient and rapid cell lysis and solubilization of proteins for both suspension and adherent cells. Lysates can be used in many downstream applications without removing the CelLytic M such as reporter gene assays Western blots/immunoprecipitation electrophoretic mobility shift assays phosphatase assays and kinase asssays.