Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLytcTM Y Cell Lyss Re500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C4482500ML
Encompass
CelLytic Y Cell Lysis Reagent is versatile phosphate-free and non-denaturing. It provides an effective method for cell lysis and protein solubilization. Target proteins maintain immunoreactivity or biological function. The protocol is brief and preformed at room temperature. No extreme conditions or glass beads are required. Use only 2.5-5 ml/gram yeast cells.
Catalog No. 50-290-6183