Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLytic M, 50 mL
Catalog No. NC0579399
Encompass
CelLytic M is a proprietary detergent solution designed for efficient whole-cell protein extraction from cultured mammalian cells. It enables efficient and rapid cell lysis and solubilization of proteins for both suspension and adherent cells. Lysates can be used in many downstream applications without removing the CelLytic M such as reporter gene assays, Western blots/immunoprecipitation, electrophoretic mobility shift assays, phosphatase assays and kinase asssays.
Catalog No. NC0579399 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C297850ML
CelLytic M is a proprietary detergent solution designed for efficient whole-cell protein extraction from cultured mammalian cells. It enables efficient and rapid cell lysis and solubilization of proteins for both suspension and adherent cells. Lysates can be used in many downstream applications without removing the CelLytic M such as reporter gene assays, Western blots/immunoprecipitation, electrophoretic mobility shift assays, phosphatase assays and kinase asssays.