Learn More
Sigma Aldrich Fine Chemicals Biosciences CelLyticTM B Plus Kit for
Catalog No. 502905751
Encompass
The CelLytic(TM) B Plus Kit is designed to efficiently lyse cells and extract proteins from both Gram negative and difficult to lyse Gram positive bacteria. This is accomplished using the standard CelLytic B a proprietary non-ionic detergent in concert with lysozyme Benzonase(R) and protease inhibitors. This complete kit takes the guesswork out of protein extraction from a variety of bacterial species.
Catalog No. 50-290-5751 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CB00501KT
The CelLytic(TM) B Plus Kit is designed to efficiently lyse cells and extract proteins from both Gram negative and difficult to lyse Gram positive bacteria. This is accomplished using the standard CelLytic B a proprietary non-ionic detergent in concert with lysozyme Benzonase(R) and protease inhibitors. This complete kit takes the guesswork out of protein extraction from a variety of bacterial species.